Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pik3r4
Entrez Gene ID
75669
Gene Name
phosphatidylinositol 3 kinase, regulatory subunit, polypeptide 4, p150
Gene Aliases
2210010O15Rik, C730038E05Rik, C85833, D9Ertd418e
Chromosome Location
Chr.9: 105642995 - 105687655 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081309.1 NP_001074778.1 18 - 19 4234 70
GenBank mRNA AK008703.1 - 3 - 4 236 70
AK087938.1 - 18 - 19 4225 70
BC017537.1 - 9 - 10 2124 70
BC023047.1 - 7 - 8 883 70
BC024083.1 - 15 - 16 2708 70
BC092149.1 - 2 - 3 193 70
BC157947.1 - 18 - 19 4059 70

View Full Product Details