Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dus3l
Entrez Gene ID
224907
Gene Name
dihydrouridine synthase 3-like (S. cerevisiae)
Gene Aliases
AI662135, AW557805
Chromosome Location
Chr.17: 56764717 - 56770093 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144858.2 NP_659107.2 3 - 4 968 64
GenBank mRNA AK157702.1 - 3 - 4 909 64
AK164614.1 - 2 - 3 568 64
AK166183.1 - 3 - 4 862 64
AY040840.1 - 3 - 4 941 64
BC052481.1 - 3 - 4 935 64
BC053941.1 - 3 - 4 935 64
BC054069.1 - 3 - 4 935 64
CK387816.1 - 1 - 2 255 64

View Full Product Details