Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl15
Entrez Gene ID
236904
Gene Name
kelch-like 15
Gene Aliases
6330500C13Rik
Chromosome Location
Chr.X: 94234495 - 94277724 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039059.1 NP_001034148.1 2 - 3 402 89
NM_001039061.1 NP_001034150.1 1 - 2 497 89
GenBank mRNA AK078112.1 - 1 - 2 438 89

View Full Product Details