The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027908.1 | NP_082184.1 | 8 - 9 | 1251 | 83 |
GenBank mRNA | AK005066.1 | - | 8 - 9 | 1207 | 83 |
AK090143.1 | - | 8 - 9 | 1242 | 83 | |
BC022626.1 | - | 8 - 9 | 1129 | 83 | |
BC043681.1 | - | 8 - 9 | 1184 | 83 | |
BC055025.1 | - | 8 - 9 | 1149 | 83 | |
BC118616.1 | - | 8 - 9 | 1238 | 83 | |
BC121826.1 | - | 8 - 9 | 1237 | 83 |