Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr43
Entrez Gene ID
72515
Gene Name
WD repeat domain 43
Gene Aliases
2610318G08Rik, AU020887, AV024208, Wrd43, mKIAA0007
Chromosome Location
Chr.17: 71615900 - 71659031 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175639.1 NP_783570.1 15 - 16 1769 83
GenBank mRNA AK012043.1 - 15 - 16 1754 83
AK032907.1 - 15 - 16 1743 83
AK129031.1 - 15 - 16 1687 83
AK152321.1 - 15 - 16 1759 83
BC079913.1 - 14 - 15 1470 83
BC117724.2 - 16 - 17 1970 83

View Full Product Details