Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nhsl2
Entrez Gene ID
100042480
Gene Name
NHS-like 2
Gene Aliases
1110062M06Rik, 6430511F03, AI225852, AW488865, EG621083, ENSMUSG00000073037, Gm10456
Chromosome Location
Chr.X: 101848669 - 102092055 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163610.1 NP_001157082.1 6 - 7 3386 70

View Full Product Details