The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008451.2 | NP_032477.2 | 3 - 4 | 760 | 70 |
NR_045528.1 | 3 - 4 | 797 | 70 | ||
GenBank mRNA | AK141439.1 | - | 3 - 4 | 690 | 70 |
BC014845.1 | - | 3 - 4 | 604 | 70 | |
BQ887879.1 | - | 3 - 4 | 628 | 70 |