Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bbs1
Entrez Gene ID
52028
Gene Name
Bardet-Biedl syndrome 1 (human)
Gene Aliases
AI451249, D19Ertd609e
Chromosome Location
Chr.19: 4886878 - 4906627 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033128.3 NP_001028300.1 1 - 2 73 78
GenBank mRNA AK037753.1 - 1 - 2 59 78
AK045850.1 - 1 - 2 70 78
AK161343.1 - 1 - 2 67 78
AK168901.1 - 1 - 2 74 78
BC076577.1 - 1 - 2 62 78
BY063920.1 - 1 - 2 74 78

View Full Product Details