Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Npas2
Entrez Gene ID
18143
Gene Name
neuronal PAS domain protein 2
Gene Aliases
MOP4, bHLHe9
Chromosome Location
Chr.1: 39193956 - 39363240 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008719.2 NP_032745.2 2 - 3 532 91
GenBank mRNA BC109166.2 - 2 - 3 532 91
U77969.1 - 2 - 3 606 91

View Full Product Details