Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Edn3
Entrez Gene ID
13616
Gene Name
endothelin 3
Gene Aliases
114CH19, ET-3, PPET3, ls, tmgc48
Chromosome Location
Chr.2: 174760758 - 174784042 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007903.4 NP_031929.1 4 - 5 820 78
GenBank mRNA AK046164.1 - 3 - 4 782 78
AK048006.1 - 4 - 5 743 78
AK049775.1 - 4 - 5 806 78
AK079150.1 - 4 - 5 819 78
AK162475.1 - 3 - 4 1021 78
BC137727.1 - 4 - 5 756 78
BC137730.1 - 4 - 5 756 78
U32330.1 - 4 - 5 960 78

View Full Product Details