The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020046.3 | NP_064430.1 | 1 - 2 | 322 | 74 |
GenBank mRNA | AK013102.1 | - | 1 - 2 | 274 | 74 |
AK088863.1 | - | 1 - 2 | 44 | 74 | |
AK149350.1 | - | 1 - 2 | 70 | 74 | |
BC027829.1 | - | 1 - 2 | 283 | 74 | |
BC045206.1 | - | 1 - 2 | 272 | 74 |