Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfhx3
Entrez Gene ID
11906
Gene Name
zinc finger homeobox 3
Gene Aliases
A230102L03Rik, Atbf1, WBP9, mKIAA4228
Chromosome Location
Chr.8: 108704034 - 108961636 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007496.2 NP_031522.2 1 - 2 685 79
GenBank mRNA D26046.1 - 1 - 2 682 79

View Full Product Details