The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199303.2 | NP_955007.2 | 14 - 15 | 1325 | 77 |
GenBank mRNA | AK087711.1 | - | 3 - 4 | 179 | 77 |