Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tm9sf4
Entrez Gene ID
99237
Gene Name
transmembrane 9 superfamily protein member 4
Gene Aliases
AA986553, AU045326, B930079E06, mKIAA0255
Chromosome Location
Chr.2: 153161301 - 153210466 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133847.3 NP_598608.2 11 - 12 1424 59
GenBank mRNA AB093220.2 - 10 - 11 1168 59
AK035622.1 - 11 - 12 1386 59
AK048158.1 - 11 - 12 1182 59
AK083541.1 - 11 - 12 1424 59
AK087048.1 - 11 - 12 1177 59
AK088012.1 - 11 - 12 1180 59
AK162435.1 - 11 - 12 1185 59
AK171487.1 - 11 - 12 1182 59
AK210804.1 - 2 - 3 147 59
BC110309.1 - 11 - 12 1180 59
BC116320.2 - 11 - 12 1376 59

View Full Product Details