The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206851.1 | NP_001193780.1 | 6 - 7 | 2176 | 72 |
GenBank mRNA | AK041893.1 | - | 1 - 2 | 157 | 72 |