Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nr1h4
Entrez Gene ID
20186
Gene Name
nuclear receptor subfamily 1, group H, member 4
Gene Aliases
AI957360, Fxr, HRR1, RIP14, Rxrip14
Chromosome Location
Chr.10: 89454234 - 89533645 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163504.1 NP_001156976.1 6 - 7 1116 104
NM_001163700.1 NP_001157172.1 5 - 6 912 104
NM_009108.2 NP_033134.2 5 - 6 900 104
GenBank mRNA AK002513.1 - 5 - 6 898 104
AK132255.1 - 5 - 6 892 104
BC015261.1 - 5 - 6 871 104
U09417.1 - 6 - 7 1080 104

View Full Product Details