Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Taf1c
Entrez Gene ID
21341
Gene Name
TATA box binding protein (Tbp)-associated factor, RNA polymerase I, C
Gene Aliases
Tafi95, mTAFI95
Chromosome Location
Chr.8: 119597974 - 119605240 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021441.2 NP_067416.2 13 - 14 1592 114
GenBank mRNA AK052250.1 - 13 - 14 1632 114
AK087534.1 - 13 - 14 1582 114
AK154846.1 - 13 - 14 1576 114
AK156718.1 - 13 - 14 1559 114
BC051944.1 - 13 - 14 1644 114
BC058383.1 - 13 - 14 1534 114
Y09974.1 - 13 - 14 1719 114

View Full Product Details