Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc40
Entrez Gene ID
207607
Gene Name
coiled-coil domain containing 40
Gene Aliases
B930008I02Rik
Chromosome Location
Chr.11: 119228572 - 119265248 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175430.4 NP_780639.1 2 - 3 1404 79
GenBank mRNA AK046971.1 - 2 - 3 1407 79
AK054172.1 - 3 - 4 918 79
BC072591.1 - 4 - 5 1169 79
BC085159.1 - 4 - 5 1164 79

View Full Product Details