The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166506.1 | NP_001159978.1 | 13 - 14 | 1818 | 58 |
NM_001166507.1 | NP_001159979.1 | 13 - 14 | 1818 | 58 | |
NM_028777.3 | NP_083053.2 | 13 - 14 | 1818 | 58 | |
GenBank mRNA | AK004818.1 | - | 13 - 14 | 1743 | 58 |
AK172947.1 | - | 10 - 11 | 1353 | 58 | |
AK195036.1 | - | 1 - 2 | 128 | 58 | |
AK200242.1 | - | 1 - 2 | 128 | 58 | |
BC005766.1 | - | 13 - 14 | 1776 | 58 | |
BC086620.1 | - | 13 - 14 | 1751 | 58 |