Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxo39
Entrez Gene ID
628100
Gene Name
F-box protein 39
Gene Aliases
1700010H23Rik
Chromosome Location
Chr.11: 72314370 - 72319419 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099688.2 NP_001093158.1 2 - 3 1196 78

View Full Product Details