The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128086.2 | NP_001121558.2 | 18 - 19 | 3034 | 74 |
NM_001177396.1 | NP_001170867.1 | 4 - 5 | 865 | 74 | |
NM_001177397.1 | NP_001170868.1 | 4 - 5 | 480 | 74 | |
GenBank mRNA | AK032061.1 | - | 2 - 3 | 136 | 74 |
AK034154.1 | - | 5 - 6 | 500 | 74 | |
AK038609.1 | - | 2 - 3 | 364 | 74 | |
AK132228.1 | - | 4 - 5 | 832 | 74 | |
BC098373.1 | - | 4 - 5 | 480 | 74 |