Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zmynd8
Entrez Gene ID
228880
Gene Name
zinc finger, MYND-type containing 8
Gene Aliases
1110013E22Rik, 2010005I16Rik, 3632413B07Rik, AI316811, AL024039, Prkcbp1, RACK7
Chromosome Location
Chr.2: 165784152 - 165898984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252584.2 NP_001239513.1 20 - 21 3445 95
NM_001252585.1 NP_001239514.1 19 - 20 3337 95
NM_001281926.1 NP_001268855.1 20 - 21 3344 95
NM_001291158.1 NP_001278087.1 21 - 22 5610 95
NM_027230.5 NP_081506.3 21 - 22 3688 95
GenBank mRNA AB093284.2 - 21 - 22 5602 95
AK014397.1 - 8 - 9 1745 95
AK132427.1 - 19 - 20 3339 95
AK147620.1 - 20 - 21 3345 95
AK155864.1 - 20 - 21 3250 95
BC041797.1 - 10 - 11 2416 95
BC048186.1 - 21 - 22 3495 95

View Full Product Details