Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zmynd8
Entrez Gene ID
228880
Gene Name
zinc finger, MYND-type containing 8
Gene Aliases
1110013E22Rik, 2010005I16Rik, 3632413B07Rik, AI316811, AL024039, Prkcbp1, RACK7
Chromosome Location
Chr.2: 165784152 - 165898984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252584.2 NP_001239513.1 21 - 22 3513 109
NM_001252585.1 NP_001239514.1 20 - 21 3405 109
NM_001281926.1 NP_001268855.1 21 - 22 3412 109
NM_001291158.1 NP_001278087.1 22 - 23 5678 109
NM_027230.5 NP_081506.3 22 - 23 3756 109
GenBank mRNA AB093284.2 - 22 - 23 5670 109
AK014397.1 - 9 - 10 1813 109
AK132427.1 - 20 - 21 3407 109
AK147620.1 - 21 - 22 3413 109
AK155864.1 - 21 - 22 3318 109
BC041797.1 - 11 - 12 2484 109
BC048186.1 - 22 - 23 3563 109

View Full Product Details