The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252584.2 | NP_001239513.1 | 8 - 9 | 951 | 82 |
NM_001252585.1 | NP_001239514.1 | 7 - 8 | 843 | 82 | |
NM_001252587.2 | NP_001239516.1 | 9 - 10 | 963 | 82 | |
NM_001281926.1 | NP_001268855.1 | 8 - 9 | 850 | 82 | |
NM_001291158.1 | NP_001278087.1 | 9 - 10 | 2960 | 82 | |
NM_027230.5 | NP_081506.3 | 9 - 10 | 1038 | 82 | |
GenBank mRNA | AB093284.2 | - | 9 - 10 | 2952 | 82 |
AK132427.1 | - | 7 - 8 | 845 | 82 | |
AK147620.1 | - | 8 - 9 | 851 | 82 | |
AK155864.1 | - | 8 - 9 | 756 | 82 | |
BC023300.1 | - | 8 - 9 | 818 | 82 | |
BC048186.1 | - | 9 - 10 | 845 | 82 | |
BC063267.1 | - | 8 - 9 | 744 | 82 | |
CA328656.1 | - | 5 - 6 | 468 | 82 | |
CK634740.1 | - | 4 - 5 | 330 | 82 | |
CO428932.1 | - | 6 - 7 | 607 | 82 |