The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252448.1 | NP_001239377.1 | 4 - 5 | 305 | 66 |
GenBank mRNA | AK138358.1 | - | 4 - 5 | 306 | 66 |
BC052063.1 | - | - | 325 | 66 |