The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252448.1 | NP_001239377.1 | 7 - 8 | 637 | 88 |
NM_001252449.1 | NP_001239378.1 | 5 - 6 | 526 | 88 | |
NM_028126.3 | NP_082402.1 | 5 - 6 | 526 | 88 | |
GenBank mRNA | AK011451.1 | - | 5 - 6 | 472 | 88 |
AK138358.1 | - | 7 - 8 | 638 | 88 | |
AY341872.1 | - | 5 - 6 | 485 | 88 | |
AY341878.1 | - | 5 - 6 | 485 | 88 | |
BC058517.1 | - | 5 - 6 | 427 | 88 |