The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172397.3 | NP_765985.1 | 2 - 3 | 183 | 113 |
GenBank mRNA | AK012581.1 | - | 2 - 3 | 157 | 113 |
AK032430.1 | - | 2 - 3 | 184 | 113 | |
AK040486.1 | - | 2 - 3 | 184 | 113 | |
AK049809.1 | - | 2 - 3 | 153 | 113 | |
AK167349.1 | - | 2 - 3 | 153 | 113 | |
AK169829.1 | - | 2 - 3 | 154 | 113 | |
AK171794.1 | - | 2 - 3 | 462 | 113 | |
BC068130.1 | - | 2 - 3 | 127 | 113 |