The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026922.1 | NP_081198.1 | 26 - 27 | 2872 | 95 |
GenBank mRNA | AK007419.1 | - | 3 - 4 | 348 | 95 |
BC026636.1 | - | 10 - 11 | 1190 | 95 | |
BC037514.1 | - | 1 - 2 | 153 | 95 | |
BC042772.1 | - | 17 - 18 | 1890 | 95 | |
EF688288.1 | - | 26 - 27 | 2716 | 95 |