Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ltf
Entrez Gene ID
17002
Gene Name
lactotransferrin
Gene Aliases
Csp82, Lf, MMS10R, Ms10r
Chromosome Location
Chr.9: 111019292 - 111042766 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008522.3 NP_032548.2 4 - 5 533 66
GenBank mRNA AK036491.1 - 4 - 5 534 66
AK144556.1 - 4 - 5 534 66
AK150799.1 - 4 - 5 534 66
AK151822.1 - 4 - 5 534 66
AK164781.1 - 4 - 5 535 66
AY792500.1 - 4 - 5 499 66
BC006904.1 - 4 - 5 541 66
BC008530.1 - 4 - 5 514 66
CT010339.1 - 4 - 5 499 66
D88510.1 - 4 - 5 533 66
FJ538998.1 - 4 - 5 499 66
J03298.1 - 4 - 5 502 66

View Full Product Details