Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il34
Entrez Gene ID
76527
Gene Name
interleukin 34
Gene Aliases
2010004A03Rik, AI593503
Chromosome Location
Chr.8: 110741829 - 110805950 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135100.2 NP_001128572.1 2 - 3 603 74
NM_029646.3 NP_083922.1 2 - 3 603 74
GenBank mRNA AK008082.1 - 2 - 3 551 74
BC016254.1 - 2 - 3 582 74

View Full Product Details