Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fanca
Entrez Gene ID
14087
Gene Name
Fanconi anemia, complementation group A
Gene Aliases
AW208693, FACA
Chromosome Location
Chr.8: 123268244 - 123318651 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016925.3 NP_058621.2 34 - 35 3411 78
GenBank mRNA AF178934.1 - 34 - 35 3407 78
AF208116.1 - 34 - 35 3413 78
AF208120.1 - 13 - 14 1494 78
BC150705.1 - 34 - 35 3409 78

View Full Product Details