Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tango6
Entrez Gene ID
272538
Gene Name
transport and golgi organization 6
Gene Aliases
AW413431, E330010G16, Tmco7
Chromosome Location
Chr.8: 106683068 - 106851439 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173037.1 NP_766625.1 10 - 11 1894 61
GenBank mRNA AK047206.1 - 5 - 6 612 61
AK054283.1 - 10 - 11 1848 61
AK077757.1 - 10 - 11 1879 61
AK085030.1 - 10 - 11 1894 61
AK173250.1 - 10 - 11 1801 61
BC066134.1 - 10 - 11 1824 61
BC066869.1 - 10 - 11 1824 61
BC069838.1 - 10 - 11 1824 61

View Full Product Details