Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tango6
Entrez Gene ID
272538
Gene Name
transport and golgi organization 6
Gene Aliases
AW413431, E330010G16, Tmco7
Chromosome Location
Chr.8: 106683068 - 106851439 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173037.1 NP_766625.1 2 - 3 829 117
GenBank mRNA AK054283.1 - 2 - 3 117
AK076373.1 - 2 - 3 752 117
AK077757.1 - 2 - 3 815 117
AK085030.1 - 2 - 3 829 117
AK173250.1 - 2 - 3 736 117
BC066134.1 - 2 - 3 759 117
BC066869.1 - 2 - 3 759 117
BC069838.1 - 2 - 3 759 117

View Full Product Details