Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cklf
- Entrez Gene ID
-
75458
- Gene Name
- chemokine-like factor
- Gene Aliases
-
1700001C14Rik, 1810018M11Rik, C32, CKLF1, CKLF3, CKLF4, CKLF5, Cklf2, Cklf6, HSPC224, UCK-1
- Chromosome Location
-
Chr.8: 104250861 - 104264936 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK156237.1 |
- |
|
638 |
|
Chr. - - [-] on Build |
86 |
View Full Product Details