Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr59
Entrez Gene ID
319481
Gene Name
WD repeat domain 59
Gene Aliases
5430401O09Rik, Cdw12
Chromosome Location
Chr.8: 111448781 - 111522107 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170742.1 NP_001164213.1 16 - 17 1766 84
NM_001170743.1 NP_001164214.1 16 - 17 1766 84
NM_176923.4 NP_795897.2 16 - 17 1766 84
GenBank mRNA AK030644.1 - 16 - 17 1647 84
AK040807.1 - 1 - 2 53 84
AK136303.1 - 12 - 13 1318 84
AK170910.1 - 16 - 17 1765 84
AK173301.1 - 16 - 17 1729 84
BC066082.1 - 16 - 17 1751 84
EF011622.1 - 16 - 17 1751 84

View Full Product Details