Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam65a
Entrez Gene ID
75687
Gene Name
family with sequence similarity 65, member A
Gene Aliases
2310066E14Rik, AI837433
Chromosome Location
Chr.8: 105594087 - 105622219 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081241.2 NP_001074710.2 11 - 12 1087 82
GenBank mRNA AK052316.1 - 11 - 12 1036 82
AK122573.1 - 11 - 12 1061 82
AK188393.1 - 2 - 3 230 82
BC030451.1 - 1 - 2 150 82
BC079880.1 - 11 - 12 1001 82

View Full Product Details