Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cfdp1
Entrez Gene ID
23837
Gene Name
craniofacial development protein 1
Gene Aliases
AA408409, Bcnt, Bucentaur, Cfdp, cp27
Chromosome Location
Chr.8: 111768473 - 111854310 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011801.1 NP_035931.1 2 - 3 283 79
GenBank mRNA AB010828.1 - 2 - 3 283 79
AK133989.1 - 2 - 3 301 79
AK160532.1 - 2 - 3 248 79
BC005589.1 - 2 - 3 264 79
Y08219.1 - 2 - 3 282 79

View Full Product Details