Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cfdp1
Entrez Gene ID
23837
Gene Name
craniofacial development protein 1
Gene Aliases
AA408409, Bcnt, Bucentaur, Cfdp, cp27
Chromosome Location
Chr.8: 111768473 - 111854310 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011801.1 NP_035931.1 6 - 7 905 63
GenBank mRNA AB010828.1 - 6 - 7 905 63
AK133989.1 - 6 - 7 923 63
AK160532.1 - 6 - 7 870 63
BC005589.1 - 6 - 7 886 63

View Full Product Details