The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007788.3 | NP_031814.2 | 4 - 5 | 578 | 100 |
GenBank mRNA | AK011501.1 | - | 4 - 5 | 484 | 100 |
AK031617.1 | - | 4 - 5 | 543 | 100 | |
AK146032.1 | - | 4 - 5 | 537 | 100 | |
AK158077.1 | - | 4 - 5 | 508 | 100 | |
BC026149.1 | - | 4 - 5 | 482 | 100 | |
BC060742.1 | - | 4 - 5 | 533 | 100 | |
BC089343.1 | - | 4 - 5 | 456 | 100 | |
U17112.1 | - | 3 - 4 | 317 | 100 | |
U51866.1 | - | 4 - 5 | 520 | 100 |