Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d20
Entrez Gene ID
67231
Gene Name
TBC1 domain family, member 20
Gene Aliases
1110028I04Rik, 2810442O16Rik, AI414693
Chromosome Location
Chr.2: 152293872 - 152312590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024196.3 NP_077158.1 3 - 4 437 70
GenBank mRNA AK006877.1 - 3 - 4 434 70
AK036751.1 - - 347 70
AK076124.1 - 3 - 4 438 70
AK082369.1 - - 433 70
AK085702.1 - - 435 70
AK158274.1 - - 434 70
AK168985.1 - - 438 70
AK172349.1 - - 434 70
BC002196.1 - 2 - 3 253 70
BC034504.1 - - 419 70
BC040089.1 - 3 - 4 400 70
BC086617.1 - - 69

View Full Product Details