Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rspry1
Entrez Gene ID
67610
Gene Name
ring finger and SPRY domain containing 1
Gene Aliases
4930470D19Rik, AI608258
Chromosome Location
Chr.8: 94601941 - 94660277 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026274.4 NP_080550.3 10 - 11 1802 61
GenBank mRNA AK031153.1 - 1 - 2 118 61
AK032416.1 - 10 - 11 1765 61
AK076799.1 - 10 - 11 1375 61
AK134544.1 - 7 - 8 669 61
AK141443.1 - 10 - 11 1352 61
AK150941.1 - 9 - 10 1277 61
AK152435.1 - 9 - 10 1277 61
AK172567.1 - 10 - 11 1768 61
AK173313.1 - 10 - 11 1421 61
BC010833.1 - 10 - 11 1417 61
BC054121.1 - 10 - 11 1788 61

View Full Product Details