The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008609.3 | NP_032635.1 | 9 - 10 | 2093 | 75 |
GenBank mRNA | BC047278.1 | - | 9 - 10 | 2093 | 75 |
BC057952.1 | - | 9 - 10 | 1663 | 75 | |
D86332.1 | - | 9 - 10 | 1589 | 75 |