Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnaic2
Entrez Gene ID
432611
Gene Name
dynein, axonemal, intermediate chain 2
Gene Aliases
C030015H18Rik, Dnai2, Gm728
Chromosome Location
Chr.11: 114722210 - 114757889 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034878.2 NP_001030050.2 12 - 13 2102 83
GenBank mRNA EF565270.1 - 11 - 12 1718 83

View Full Product Details