Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnaic2
Entrez Gene ID
432611
Gene Name
dynein, axonemal, intermediate chain 2
Gene Aliases
C030015H18Rik, Dnai2, Gm728
Chromosome Location
Chr.11: 114722210 - 114757889 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034878.2 NP_001030050.2 2 - 3 573 64
GenBank mRNA AK138639.1 - 2 - 3 500 64
EF565270.1 - 1 - 2 189 64

View Full Product Details