Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cog1
Entrez Gene ID
16834
Gene Name
component of oligomeric golgi complex 1
Gene Aliases
6030445I15, Ldlb, mKIAA1381
Chromosome Location
Chr.11: 113649178 - 113669331 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013581.3 NP_038609.3 4 - 5 937 92
GenBank mRNA AK031510.1 - 3 - 4 393 92
AK034636.2 - 4 - 5 942 92
AK152464.1 - 4 - 5 928 92
AK158507.1 - 4 - 5 1299 92
AK159497.1 - 4 - 5 939 92
AK162971.1 - 4 - 5 938 92
AK171574.1 - 4 - 5 931 92
AK207477.1 - 2 - 3 196 92
AK220387.1 - 4 - 5 934 92
BC063056.1 - 4 - 5 919 92

View Full Product Details