The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026682.2 | NP_080958.2 | 1 - 2 | 138 | 85 |
GenBank mRNA | AK161074.1 | - | 1 - 2 | 139 | 85 |
BC016190.1 | - | 1 - 2 | 132 | 85 | |
DV659633.1 | - | 1 - 2 | 128 | 85 |