Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
D11Wsu47e
Entrez Gene ID
276852
Gene Name
DNA segment, Chr 11, Wayne State University 47, expressed
Gene Aliases
-
Chromosome Location
Chr.11: 113684400 - 113727444 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK017508.1 - 1 - 2 260 113
AK171307.1 - 1 - 2 303 113

View Full Product Details