Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ubp1
Entrez Gene ID
22221
Gene Name
upstream binding protein 1
Gene Aliases
Cp2b, LBP-1a, LBP-1b, NF2d9
Chromosome Location
Chr.9: 113930934 - 113977202 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083319.1 NP_001076788.1 2 - 3 527 86
NM_013699.2 NP_038727.1 2 - 3 527 86
GenBank mRNA AB221581.1 - 1 - 2 116 86
AK143282.1 - 2 - 3 528 86
AY182062.1 - 1 - 2 116 86
BC034749.1 - 2 - 3 216 86
BC139828.1 - 1 - 2 144 86
BC141082.1 - 1 - 2 206 86
U20086.1 - 1 - 2 118 86

View Full Product Details