Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ubp1
Entrez Gene ID
22221
Gene Name
upstream binding protein 1
Gene Aliases
Cp2b, LBP-1a, LBP-1b, NF2d9
Chromosome Location
Chr.9: 113930934 - 113977202 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083319.1 NP_001076788.1 12 - 13 1597 136
NM_013699.2 NP_038727.1 11 - 12 1489 136
GenBank mRNA AB221581.1 - 10 - 11 1077 136
AK079815.1 - 4 - 5 324 136
AK140942.1 - 6 - 7 494 136
AY182062.1 - 11 - 12 1186 136
BC034749.1 - 10 - 11 1816 136
BC139828.1 - 10 - 11 1106 136
BC141082.1 - 10 - 11 1168 136
U20086.1 - 10 - 11 1080 136

View Full Product Details