Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxw27
Entrez Gene ID
76998
Gene Name
F-box and WD-40 domain protein 27
Gene Aliases
1700124P09Rik
Chromosome Location
Chr.9: 109765655 - 109790653 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BB702760.1 - 1 - 2 89 81

View Full Product Details